Vanadium ≥99.8% (metals basis), foil, Thickness 0.075 mm (0.003 in)

Supplier: Thermo Scientific Chemicals

44106-GK 44106-GK 44106-FY 44106-FY
AA44106-GKEA 743.99 USD
CAAA44106-FY AA44106-GK CAAA44106-GK AA44106-FY
Vanadium ≥99.8% (metals basis), foil, Thickness 0.075 mm (0.003 in)
Vanadium
Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477 °C (1013 hPa)
Melting Pt: 1896 °C
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR