Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

043219-FI 043219-FF
AA43219-FIEA 918 USD
AA43219-FI AA43219-FF
Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.1 mm (0.004 in)
Scandium

MDL: MFCD00016323


Purity: 99.9% CAS No.: 7440-20-2 Physical Form: ≈0.20g/25x25mm, Packaged under argon Odor: Odorless MDL No.: MFCD00016323

Formula: Sc
MW: 44.96 g/mol
Boiling Pt: 2730
Melting Pt: 1541
Density: 2.989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR