Glassy carbon, rods, Diameter 4 mm (0.16 in) Type 1

Supplier: Thermo Scientific Chemicals

042823-DM 042823-DJ 042823-DR
AA42823-DMEA 454 USD
AA42823-DM AA42823-DJ AA42823-DR
Glassy carbon, rods, Diameter 4 mm (0.16 in) Type 1
Glassy carbon

MDL: MFCD00133992


Physical Form: ≈0.19g/cm MDL No.: MFCD00133992

Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR