Vanadium ≥99.7% (metals basis), pieces

Supplier: Thermo Scientific Chemicals

042775-30 042775-18 042775-09
AA42775-30EA 761 USD
AA42775-30 AA42775-18 AA42775-09
Vanadium ≥99.7% (metals basis), pieces
Vanadium

MDL: MFCD00011453


Purity: 99.7% CAS No.: 7440-62-2 Odor: Odorless MDL No.: MFCD00011453

Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR