Deuterium chloride 20% (w/w) in D₂O (99.96% D)

Supplier: Thermo Scientific Chemicals
Danger

042406-06
AA42406-06EA 254 USD
AA42406-06
Deuterium chloride 20% (w/w) in D₂O (99.96% D)
Deuterium chloride
Formula: DCl
MW: 37.45 g/mol
Boiling Pt: 108 °C
Density: 1.20
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR