Dichloromethane-[D₂] (100% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methylene chloride-D2
42334-KV
42334-SA
42334-SA
042334-KV
AA42334-KVEA
725
USD
CAAA42334-KV
AA42334-KV
CAAA42334-SA
AA42334-SA
Dichloromethane-[D₂] (100% D) (Isotopic) for NMR spectroscopy
Dichloromethane-[D2]
Formula:
CD₂Cl₂ MW: 86.92 g/mol Boiling Pt: 40 °C (1013 hPa) Melting Pt: –97 °C Density: 1.35 Storage Temperature: Ambient |
MDL Number:
MFCD00000882 CAS Number: 1665-00-5 EINECS: 216-776-0 UN: 1593 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...