Tetrahydrofuran, anhydrous 99,8 Unstabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide
Applications: Synthesis and Liquid Chromotography
Purity: 99.8+%
CAS No.: 109-99-9
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -17°(1°F)
Physical Form: Liquid
Density: 0.889
Refractive Index: 1.4070
Appearance: Colorless
Odor: Ether-like
Melting Point: -108°
Boiling Point: 66°
Beilstein Reference: 102391
Merck Reference: 14,9211
UN No.: 2056
MDL No.: MFCD00005356
Synthesis and Liquid Chromotography
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 66 Melting Pt: -108.5 Density: 0.889 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3 II REACH: 05-2114615188-48 Merck Index: 14,9211 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...