Boron ≥99.4% (metals basis), crystalline pieces 1-30 mm (0.039-1.2 in)

Supplier: Thermo Scientific Chemicals
Warning

041858-30 041858-09 041858-18
AA41858-30EA 3557 USD
AA41858-30 AA41858-09 AA41858-18
Boron ≥99.4% (metals basis), crystalline pieces 1-30 mm (0.039-1.2 in)
Boron

MDL: MFCD00134034


Purity: 99.4% CAS No.: 7440-42-8 Odor: Odorless MDL No.: MFCD00134034

Formula: B
Density: 1.73 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR