Bismuth ≥99.999% (metals basis), foil, Thickness 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

041637-FI 41637-FF 041637-FF 41637-FI
AA41637-FIEA 2117.67 USD
AA41637-FI AA41637-FF CAAA41637-FI CAAA41637-FF
Bismuth ≥99.999% (metals basis), foil, Thickness 2.0 mm (0.08 in)
Bismuth
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR