Boron carbide ≥99.5% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

041050-KS 41050-KS
AA41050-KSEA 1391 USD
AA41050-KS CAAA41050-KS
Boron carbide ≥99.5% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)
Boron carbide
Formula: CB₄
Boiling Pt: ∼3500 °C (1013 hPa)
Melting Pt: 2350 °C
Density: 2.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011520
CAS Number: 12069-32-8
EINECS: 235-111-5
Merck Index: 12,01374

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR