n-Heptane ≥96%, Environmental Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Pesticide residue, environmental, and GC analysis
Purity: 96+%
CAS No.: 142-82-5
Molecular Formula: CH3(CH2)5CH3
Formula Weight: 100.21
Flash Point: -4°(24°F)
Physical Form: Liquid
Density: 0.684
Refractive Index: 1.3877
Odor: Petroleum-like
Melting Point: -91°
Boiling Point: 97-99°
Beilstein Reference: 1730763
Merck Reference: 14,4659
UN No.: 1206
MDL No.: MFCD00009544
Pesticide residue, environmental, and GC analysis
Formula:
H₃C(CH₂)₅CH₃ Boiling Pt: 97…99 °C Melting Pt: –91 °C Density: 0.684 Flash Pt: –4 °C (24 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II Merck Index: 14,4659 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...