Iron ≥99.99% (metals basis), foil, Thickness 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

40495-FF 040495-GH 040495-FI 40495-GH 040495-FF 40495-FI
AA40495-FIEA 260.99 USD
CAAA40495-FF AA40495-FI AA40495-GH CAAA40495-GH AA40495-FF CAAA40495-FI
Iron ≥99.99% (metals basis), foil, Thickness 0.5 mm (0.02 in)
Iron
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR