Iron ≥99.99% (metals basis), foil, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

040494-FF 40494-GH 040494-GH 040494-FI 40494-FI 40494-FF
AA40494-FIEA 165.99 USD
AA40494-FI CAAA40494-GH CAAA40494-FF AA40494-GH CAAA40494-FI AA40494-FF
Iron ≥99.99% (metals basis), foil, Thickness 0.25 mm (0.01 in)
Iron
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR