Potassium hydrogen carbonate 99.7-100.5% (dried basis) ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium bicarbonate

040195-36 040195-22
AA40195-36EA 74.82 USD
AA40195-36 AA40195-22
Potassium hydrogen carbonate 99.7-100.5% (dried basis) ACS
Potassium hydrogen carbonate

MDL: MFCD00011402 Slightly soluble in water. Practically insoluble in alcohol. Fieser: 13,253


Purity: 99.7-100.5% CAS No.: 298-14-6 Molecular Formula: KHCO3 Formula Weight: 100.12 Physical Form: Granular Density: 2.17 Solubility: Slightly soluble in water. Practically insoluble in alcohol Merck Reference: 14,7609 MDL No.: MFCD00011402

Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292
Density: 2.17
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 14,7609

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.1%
PO5 5 ppm
SO3 0.003%
NH3 5 ppm max.
Ca 0.002%
MG 0.001%
Heavy metals (as Pb) 5 ppm
Fe 5 ppm
NA 0.03%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR