Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.62 mm (0.024 in)

Supplier: Thermo Scientific Chemicals

39695-FF 039695-FF 039695-FI 39695-FI
AA39695-FIEA 893 USD
AA39695-FI CAAA39695-FI AA39695-FF CAAA39695-FF
Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.62 mm (0.024 in)
Terbium
Formula: Tb
MW: 158.93 g/mol
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
Merck Index: 12,09300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR