Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), wire, annealed, Diameter 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
039383-BS
039383-BW
AA39383-BSEA
96
USD
AA39383-BS
AA39383-BW
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), wire, annealed, Diameter 0.25 mm (0.01 in)
Platinum Iridium alloy (90:10 w%)
MDL: MFCD02091683
Purity: 99.9+%
Molecular Formula: Pt:Ir; 90:10 wt%,
Physical Form: ≈0.11g/10cm, Temper: soft
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...