1,2-Dichloroethane 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylene chloride, Ethylene dichloride, Dichloroethane
039121-K2
039121-K7
039121-AP
039121-M1
AA39121-K2EA
189
USD
AA39121-K2
AA39121-K7
AA39121-AP
AA39121-M1
1,2-Dichloroethane 99+% ACS
1,2-Dichloroethane
MDL: MFCD00000963
Beilstein Registry No.: 605264
Purity: 99+%
CAS No.: 107-06-2
Molecular Formula: ClCH2CH2Cl
Formula Weight: 98.96
Flash Point: 15°(59°F)
Physical Form: Liquid
Density: 1.256
Refractive Index: 1.4448
Melting Point: -35°
Boiling Point: 83°
Beilstein Reference: 605264
Merck Reference: 14,3797
UN No.: 1184
MDL No.: MFCD00000963
Formula:
ClH₂CCH₂Cl MW: 98.96 g/mol Boiling Pt: 83 °C Melting Pt: –35 °C Density: 1.256 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000963 CAS Number: 107-06-2 EINECS: 203-458-1 UN: 1184 ADR: 3,II Merck Index: 14,3797 |
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.2% |
Titratable acid | 0.0003meq/g |
H2O | 0.03% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...