Potassium bromide, crystal optic disc 32 mm x 3 mm, unpolished
Supplier: Thermo Scientific Chemicals
|
Warning
|
038393-KT
38393-KT
AA38393-KTEA
53.8
USD
CAAA38393-KT
AA38393-KT
Potassium bromide, crystal optic disc 32 mm x 3 mm, unpolished
Potassium bromide
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 °C (1013 hPa) Melting Pt: 730 °C Density: 2.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 13,07701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...