Glassy carbon, rods, Diameter 1 mm (0.04 in) Type 1

Supplier: Thermo Scientific Chemicals

038009-DJ 038009-DM 038009-DR
AA38009-DJEA 165.5 USD
AA38009-DJ AA38009-DM AA38009-DR
Glassy carbon, rods, Diameter 1 mm (0.04 in) Type 1
Glassy carbon

MDL: MFCD00133992


Physical Form: ≈0.01g/cm MDL No.: MFCD00133992

Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR