Glassy carbon, rods, Diameter 5 mm (0.2 in) Type 2

Supplier: Thermo Scientific Chemicals

037998-DM 037998-DJ 037998-DR
AA37998-DMEA 493 USD
AA37998-DM AA37998-DJ AA37998-DR
Glassy carbon, rods, Diameter 5 mm (0.2 in) Type 2
Glassy carbon

MDL: MFCD00133992


Physical Form: ≈0.28g/cm MDL No.: MFCD00133992

Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: ,
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR