Potassium permanganate ≥99.0% ACS low in mercury
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Permanganic acid potassium salt
036675-22
036675-36
036675-A4
AA36675-22EA
63.22
USD
AA36675-22
AA36675-36
AA36675-A4
Potassium permanganate ≥99.0% ACS low in mercury
Potassium permanganate
MDL: MFCD00011364
Fieser: 1,942 14,267 15,273 16,283 18,301 19,277 20,315 21,362
Purity: 99.0% min
CAS No.: 7722-64-7
Molecular Formula: KMnO4
Formula Weight: 158.03
Physical Form: Crystalline
Density: 2.7
Appearance: Dark purple
Melting Point: 240° dec.
Merck Reference: 14,7655
UN No.: 1490
MDL No.: MFCD00011364
Formula:
KMnO₄ MW: 158.03 g/mol Melting Pt: 240 °C (dec.) Density: 2.7 Storage Temperature: Ambient |
MDL Number:
MFCD00011364 CAS Number: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 5.1 II Merck Index: 14,7655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...