Water, REacton® ACS, deionized
Supplier: Thermo Scientific Chemicals
MDL: MFCD00011332
CAS No.: 7732-18-5
Molecular Formula: H2O
Formula Weight: 18.02
Physical Form: Liquid
Density: 1.000
Refractive Index: 1.3330
Odor: Odorless
Melting Point: 0°
Boiling Point: 100°
Merck Reference: 14,10039
MDL No.: MFCD00011332
Formula:
H₂O Boiling Pt: 100 °C Melting Pt: 0 °C Density: 1.000 Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 14,10039 |
Specification Test Results
Specific conductance at 25° | 2.0x10 |
Silicate (as SiO2) | 0.01ppm ppm |
Heavy Metals (as Pb) | 0.01 ppm |
Substances reducing permanganate | Pass test |
Cl | 0.4 ppm |
NO3 | 0.4 ppm |
PO3 | 1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...