Acetic anhydride 97+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
036292-AP
036292-K3
036292.AE
AA36292-K3EA
234.16
USD
AA36292-K3
AAA36292-AE
AA36292-AP
Acetic anhydride 97+% ACS
Acetic anhydride
Formula:
(CH₃CO)₂O MW: 102.09 g/mol Boiling Pt: 138…140 °C Melting Pt: –73 °C Density: 1.082 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008705 CAS Number: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II REACH: 01-2119486470-36 Merck Index: 13,00057 |
Specification Test Results
Form | Liquid |
Cl | 5 ppm |
PO3 | 0.001% |
SO3 | 5 ppm |
Heavy Metals (as Pb) | 2 ppm |
Fe | 5 ppm |
Substances reducing permanganate | Pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...