Lead ≥99.5%, granules ACS

Supplier: Thermo Scientific Chemicals
Danger

036227-22 036227-36 036227-A3
AA36227-22EA 57.4 USD
AA36227-22 AA36227-36 AA36227-A3
Lead ≥99.5%, granules ACS
Lead

MDL: MFCD00134050


Purity: 99.5% min CAS No.: 7439-92-1 Appearance: Grey Odor: Odorless MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now

Specification Test Results

Antimony and tin (as Sn) 0.005%
As 1ppm ppm
Bi 5 ppm
Cu 3 ppm
Fe 0.001%
Ni 0.001%
Ag 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR