Copper ≥99.8% (metals basis), foil, annealed, Thickness 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
035818-FI
035818-GH
035818-FF
AA35818-FIEA
86.29
USD
AA35818-FI
AA35818-GH
AA35818-FF
Copper ≥99.8% (metals basis), foil, annealed, Thickness 0.05 mm (0.002 in)
Copper
MDL: MFCD00010965
Purity: 99.8%
CAS No.: 7440-50-8
Physical Form: ≈0.28g/25x25mm
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...