4-Methyl-2-pentanone ≥98.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl Isobutyl Ketone
, Isobutyl Methyl Ketone
, Methyl iso-Butyl Ketone
033346-K2
033346-AP
AA33346-K2EA
95.08
USD
AA33346-K2
AA33346-AP
4-Methyl-2-pentanone ≥98.5% ACS
4-Methyl-2-pentanone
Formula:
(CH₃)₂CHCH₂COCH₃ MW: 100.16 g/mol Boiling Pt: 117…118 °C Melting Pt: –84 °C Density: 0.801 Flash Pt: 13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008938 CAS Number: 108-10-1 EINECS: 203-550-1 UN: 1245 ADR: 3,II Merck Index: 13,05227 |
![]() |
Specification Test Results
Color (APHA) | 15 |
Evaporation residue | 0.005% |
Titratable acid | 0.002meq/g |
H2O | 0.1% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...