n-Heptane 99+% Spectrophotometry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: high purity solvents

032441-M1 032441-K2 032441-K7
AA32441-M1EA 254 USD
AA32441-M1 AA32441-K2 AA32441-K7
n-Heptane 99+% Spectrophotometry
n-Heptane

MDL: MFCD00009544 Beilstein Registry No.: 1730763 Notes: Meets ACS spectrophotometric requirements. Filtered through 0.2µ filters.


Purity: 99+% CAS No.: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 Flash Point: -4°(24°F) Physical Form: Liquid Density: 0.684 Refractive Index: 1.3877 Odor: Petroleum-like Melting Point: -91° Boiling Point: 97-99° Beilstein Reference: 1730763 Merck Reference: 14,4659 UN No.: 1206 MDL No.: MFCD00009544

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4
Melting Pt: -90.6
Density: 0.684
Flash Pt: –4 °C (24 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 14,4659
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Residual water 0.03%
Residue on Evaporation 3ppm
n-Heptane 96% (info)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR