Triethylphosphine 97%

Supplier: Thermo Scientific Chemicals
Danger

30177-14 030177-06 30177-06 030177-14
AA30177-06EA 136 USD
CAAA30177-14 AA30177-06 CAAA30177-06 AA30177-14
Triethylphosphine 97%
Triethylphosphine
Formula: C₆H₁₅P
Boiling Pt: 126…128 °C
Melting Pt: –3 °C
Density: 0.81
Storage Temperature: Ambient
MDL Number: MFCD00009256
CAS Number: 554-70-1
EINECS: 209-068-8
UN: 2845
ADR: 4.2,I
Merck Index: 14,9675

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR