n-Pentane 99 HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pentane
Filter 0,2 um
022907-K2
022907-K7
022907-M6
022907-M1
AA22907-K2EA
120.44
USD
AA22907-K2
AA22907-K7
AA22907-M6
AA22907-M1
n-Pentane 99 HPLC
n-Pentane
MDL: MFCD00009498
Notes: Filtered through 0.2µ filters.
Purity: 99% min
CAS No.: 109-66-0
Molecular Formula: CH3(CH2)3CH3
Formula Weight: 72.15
Flash Point: -49°(-57°F)
Physical Form: Liquid
Density: 0.626
Refractive Index: 1.3580
Appearance: Colorless
Melting Point: -130°
Boiling Point: 36°
Merck Reference: 14,7116
UN No.: 1265
MDL No.: MFCD00009498
Formula:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Boiling Pt: 36 °C Melting Pt: -130 Density: 0.626 Flash Pt: –49 °C (–57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3 II REACH: 01-2119459286-30 Merck Index: 14,7116 |
|
Specification Test Results
Appearance (Color) | Colorless |
Form | Liquid |
Color (APHA) | ≤10 |
Assay (GC) | ≥99.9% ( As C5 Hydrocarbons) |
Comment | Sub micron filtered |
Water Content (Karl Fischer Titration) | ≤0.02% |
Optical Absorbance at 200 nm | ≤1.0 AU |
Optical Absorbance at 210 nm | ≤0.5 AU |
Optical Absorbance at 220 nm | ≤0.15 AU |
Optical Absorbance at 245 nm | ≤0.01 AU |
Residue on Evaporation | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...