Copper (I) sulfide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
014718-18
014718-A1
014718-30
AA14718-18EA
71.8
USD
AA14718-18
AA14718-A1
AA14718-30
Copper (I) sulfide ≥99.5% (metals basis)
Copper (I) sulphide
Luminous paints, electrodes, solar cells, as catalyst
Purity: 99.5%
CAS No.: 22205-45-4
Molecular Formula: Cu2S
Formula Weight: 159.14
Physical Form: -200 Mesh Powder
Density: 5.60
Melting Point: ca 1100°
Solubility: Practically insoluble in water, acetic acid. Partially soluble in NH4OH
Merck Reference: 14,2667
MDL No.: MFCD00016062
Luminous paints, electrodes, solar cells, as catalyst
Formula:
Cu₂S Melting Pt: ∼ 1100 °C Density: 5.60 Storage Temperature: Ambient |
MDL Number:
MFCD00016062 CAS Number: 22205-45-4 EINECS: 244-842-9 Merck Index: 14,2667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...