Ammonium formate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Formic acid ammonium salt
014517-18
014517-30
AA14517-18EA
66.71
USD
AA14517-18
AA14517-30
Ammonium formate ≥98%
Ammonium formate
Purity: 98%
CAS No.: 540-69-2
Molecular Formula: CH5NO2
Formula Weight: 63.06
Physical Form: Crystalline
Density: 1.28
Appearance: White
Odor: Ammonia-like
Melting Point: ca 116°
Beilstein Reference: 3625095
Merck Reference: 14,523
MDL No.: MFCD00013103
Fieser: 1,38 15,13 18,23 20,16
Formula:
HCO₂NH₄ MW: 63.06 g/mol Melting Pt: ∼116 °C Density: 1.28 Storage Temperature: Ambient |
MDL Number:
MFCD00013103 CAS Number: 540-69-2 EINECS: 208-753-9 Merck Index: 14,523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...