Barium ≥99.9% (metals basis), crystalline dendritic solid

Supplier: Thermo Scientific Chemicals
Danger


ampouled under argon

014454-14 014454-06
AA14454-14EA 1315.64 USD
AA14454-14 AA14454-06
Barium ≥99.9% (metals basis), crystalline dendritic solid
Barium

MDL: MFCD00134031


Purity: 99.9% CAS No.: 7440-39-3 Physical Form: Ampouled under argon Appearance: Gold/grey Odor: Odorless UN No.: 1400 MDL No.: MFCD00134031

Formula: Ba
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR