Potassium permanganate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

014307-22 014307-A3 014307-36
AA14307-22EA 65 USD
AA14307-22 AA14307-A3 AA14307-36
Potassium permanganate ≥99.0% ACS
Potassium permanganate

In photography, tanning leather, purifying water, manufacturing mineral water, as an analytical lab reagent


Purity: 99.0% min CAS No.: 7722-64-7 Molecular Formula: KMnO4 Formula Weight: 158.03 Physical Form: Crystalline Density: 2.7 Appearance: Dark purple Melting Point: 240° dec. Solubility: Partially soluble in water. Decomposes in organic solvents and acids Merck Reference: 14,7655 UN No.: 1490 MDL No.: MFCD00011364


In photography, tanning leather, purifying water, manufacturing mineral water, as an analytical lab reagent

Formula: KMnO₄
Melting Pt: 240 °C (dec.)
Density: 2.7
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 14,7655

Order Now

Specification Test Results

Insoluble matter 0.2%
Chloride and chlorate (as Cl) 0.005%
SO3 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR