Sodium periodate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium metaperiodate

013798-09 13798-36 13798-22 13798-09 013798-A3 013798-22 13798-A3 013798-36
AA13798-09EA 66.73 USD
CAAA13798-22 CAAA13798-09 AA13798-09 AA13798-36 AA13798-22 CAAA13798-A3 CAAA13798-36 AA13798-A3
Sodium periodate 98%
Sodium periodate

Source of periodic acid, as an analytical reagent, and as an oxidizing agent

Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 300 °C (dec.)
Density: 3.86
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II
Merck Index: 14,8640

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR