Tin ≥99.85% (metals basis), rods, Diameter 6.35 cm (2.5 in)
Supplier: Thermo Scientific Chemicals
12601-36
012601-36
AA12601-36EA
229.99
USD
AA12601-36
CAAA12601-36
Tin ≥99.85% (metals basis), rods, Diameter 6.35 cm (2.5 in)
Tin
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...