Potassium perchlorate, anhydrous 99.0-100.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

011630-22 011630-36
AA11630-22EA 56.22 USD
AA11630-22 AA11630-36
Potassium perchlorate, anhydrous 99.0-100.5% ACS
Potassium perchlorate

For Parr Bomb Work


Purity: 99.0-100.5% CAS No.: 7778-74-7 Molecular Formula: KClO4 Formula Weight: 138.55 Physical Form: -100 Mesh Powder Density: 2.52 Melting Point: 400° dec. Solubility: Partially soluble in water. Insoluble in alcohol. Merck Reference: 14,7653 UN No.: 1489 MDL No.: MFCD00011362


For Parr Bomb Work

Formula: ClKO₄
MW: 138.55 g/mol
Melting Pt: 400 °C (dec.)
Density: 2.52
Storage Temperature: Ambient
MDL Number: MFCD00011362
CAS Number: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: 5.1,II
Merck Index: 14,7653

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 0.3%
SO3 0.001%
Ca 0.005%
Heavy Metals (as Pb) 5 ppm max.
Fe 5%
NA 0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR