Boron 90%, amorphous powder APS <1 µm (max. 5% Mg)

Supplier: Thermo Scientific Chemicals
Warning

011338-22 011338-36
AA11338-06EA 35.18 USD
AA11338-06 AA11338-22 AA11338-36 AA11338-14
Boron 90%, amorphous powder APS <1 µm (max. 5% Mg)
Boron

MDL: MFCD00134034


Purity: 90% CAS No.: 7440-42-8 Odor: Odorless UN No.: 3178 MDL No.: MFCD00134034

Formula: B
Boiling Pt: 2550 °C (1013 hPa)
Melting Pt: 2000…2075 °C
Density: 3.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 12,01373

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR