Boron ≥98% (metals basis), powder, crystalline amorphous -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
011337-09
011337-18
AA11337-09EA
126
USD
AA11337-09
AA11337-18
Boron ≥98% (metals basis), powder, crystalline amorphous -325 mesh
Boron
Purity: 98%
CAS No.: 7440-42-8
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00134034
Formula:
B Boiling Pt: 2550 °C (1013 hPa) Melting Pt: 2000…2075 °C Density: 3.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134034 CAS Number: 7440-42-8 EINECS: 231-151-2 UN: 3178 ADR: 4.1,III Merck Index: 12,01373 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...