Vanadium ≥99.5% (metals basis), plate, annealed, Thickness 3.2 mm (0.13 in)
Supplier: Thermo Scientific Chemicals
11099-FI
011099-FI
AA11099-FIEA
529.99
USD
AA11099-FI
CAAA11099-FI
Vanadium ≥99.5% (metals basis), plate, annealed, Thickness 3.2 mm (0.13 in)
Vanadium
Formula:
V MW: 50.94 g/mol Boiling Pt: 3477 °C (1013 hPa) Melting Pt: 1896 °C Density: 6.015 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 Merck Index: 12,10053 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...