Tin ≥99.9999% (metals basis), shot, Puratronic® 3 mm (0.1 in)

Supplier: Thermo Scientific Chemicals

011009-06 011009-14 011009-22
AA11009-06EA 82.2 USD
AA11009-06 AA11009-14 AA11009-22
Tin ≥99.9999% (metals basis), shot, Puratronic® 3 mm (0.1 in)
Tin

MDL: MFCD00133862


Purity: 99.9999% CAS No.: 7440-31-5 Physical Form: Teardrop Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR