Potassium nitrate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nitrate of Potash
010844-30
010844-06
010844-18
AA10844-18EA
170.76
USD
AA10844-18
AA10844-06
AA10844-30
Potassium nitrate ≥99.995% (metals basis), Puratronic®
Potassium nitrate
Formula:
KNO₃ MW: 101.1 g/mol Boiling Pt: 400 °C (dec.) Melting Pt: 334 °C Density: 2.109 Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 14,7648 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...