Bismuth ≥99.999% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

10766-14 10766-06 010766-06 10766-22 010766-22 010766-14
AA10766-22EA 440 USD
CAAA10766-14 AA10766-22 CAAA10766-22 AA10766-06 CAAA10766-06 AA10766-14
Bismuth ≥99.999% (metals basis), powder -200 mesh
Bismuth
Formula: Bi
MW: 208.98 g/mol
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR