Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)
Supplier: Thermo Scientific Chemicals
|
Warning
|
010734-30
010734-A1
010734-18
AA10734-30EA
343
USD
AA10734-30
AA10734-A1
AA10734-18
Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)
Lithium carbonate
MDL: MFCD00011084
Purity: 99.998%
CAS No.: 554-13-2
Molecular Formula: Li2CO3
Formula Weight: 73.90
Physical Form: Powder
Density: 2.11
Appearance: White
Odor: Odorless
pH: 10-11.5
Melting Point: 723°
Boiling Point: 1310° dec.
Merck Reference: 14,5527
MDL No.: MFCD00011084
Formula:
Li₂CO₃ Boiling Pt: 1310 °C (dec.) Melting Pt: 723 °C Density: 2.11 Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 14,5527 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...