Aluminium ≥99% (metals basis), spherical powder APS 17-30 µm
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum
010576-36
010576-22
010576-A3
010576-A9
AA10576-36EA
73.9
USD
AA10576-36
AA10576-22
AA10576-A3
AA10576-A9
Aluminium ≥99% (metals basis), spherical powder APS 17-30 µm
Aluminium
MDL: MFCD00134029
Purity: 99%
CAS No.: 7429-90-5
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Grey/silver
Odor: Odorless
UN No.: 1396
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 UN: 1396 ADR: 4.3 II Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...