Titanium ≥99.7% (metals basis), wire, Length 91 cm (36 in), Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Warning

10397-KJ 10397-KM 010397-KM 010397-KJ
AA10397-KMEA 738.99 USD
AA10397-KM AA10397-KJ CAAA10397-KM CAAA10397-KJ
Titanium ≥99.7% (metals basis), wire, Length 91 cm (36 in), Diameter 2.0 mm (0.08 in)
Titanium
Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4,51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR