Tantalum ≥99.95% (metals basis), foil, annealed, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

010353-FI 010353-GH 010353-GW
AA10353-FIEA 98.8 USD
AA10353-FI AA10353-GH AA10353-GW
Tantalum ≥99.95% (metals basis), foil, annealed, Thickness 0.25 mm (0.01 in)
Tantalum

MDL: MFCD00011252


Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈10.4g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR