Niobium ≥99.8% (metals basis), rods, annealed, Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

010273-BU 010273-BY 010273-BQ
AA10273-BUEA 177 USD
AA10273-BU AA10273-BY AA10273-BQ
Niobium ≥99.8% (metals basis), rods, annealed, Diameter 6.35 mm (0.25 in)
Niobium

MDL: MFCD00011126


Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈2.7g/cm, Temper: soft Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR