Lead ≥99.9% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

010223-30 010223-A1 010223-A7
AA10223-30EA 51.34 USD
AA10223-30 AA10223-A1 AA10223-A7
Lead ≥99.9% (metals basis), powder -200 mesh
Lead

MDL: MFCD00134050


Purity: 99.9% CAS No.: 7439-92-1 Odor: Odorless UN No.: 3077 MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1749 °C (1013 hPa)
Melting Pt: 327.5 °C
Density: 11.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR