Iron ≥99.5%, spherical powder APS 6-10 µm,, reduced

Supplier: Thermo Scientific Chemicals
Danger

010214-30 010214-14 010214-36 010214-95
AA10214-30EA 100 USD
AA10214-30 AA10214-14 AA10214-36 AA10214-95
Iron ≥99.5%, spherical powder APS 6-10 µm,, reduced
Iron

MDL: MFCD00010999 Notes: Approximately 98% spherical, 2% fragments


Purity: 99.5% CAS No.: 7439-89-6 Physical Form: Apparent density 2.0-3.0g/cm3 UN No.: 3089 MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1536 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR