Platinum iridium alloy (85:15 w%), wire, hard, Diameter 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
010055-BS
010055-BW
AA10055-BSEA
141
USD
AA10055-BS
AA10055-BW
Platinum iridium alloy (85:15 w%), wire, hard, Diameter 0.5 mm (0.02 in)
Platinum Iridium alloy (85:15 w%)
MDL: MFCD02091683
Molecular Formula: Pt:Ir; 85:15 wt%
Physical Form: ≈0.44g/10cm, Temper: as drawn
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...